missing translation for 'onlineSavingsMsg'
Learn More

7-Acetyl-3,4-dihydro-1,5-benzodioxepin, 96%, Thermo Scientific™

Product Code. 11431720
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22776-09-6
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.214
MDL Number MFCD00114700
InChI Key VGRSZNPWUZDPCH-UHFFFAOYSA-N
Synonym 1-3,4-dihydro-2h-1,5-benzodioxepin-7-yl ethan-1-one, 1-3,4-dihydro-2h-1,5-benzodioxepin-7-yl ethanone, 1-3,4-dihydro-2h-benzo b 1,4 dioxepin-7-yl ethanone, 1-3,4-dihydro-2h-benzo b 1,4-dioxepin-7-yl ethanone, 7-acetyl-2h,3h,4h-benzo b 1,4-dioxepin, zlchem 1110, acmc-20apc2, 3,4-trimethylenedioxyacetophenone, 7-acetyl-3,4-dihydro-1,5-benzodioxepin, 7-acetyl-3,4-dihydro-2h-1,5-benzodioxepin
PubChem CID 2775272
IUPAC Name 1-(3,4-dihydro-2H-1,5-benzodioxepin-7-yl)ethanone
SMILES CC(=O)C1=CC2=C(C=C1)OCCCO2

Specifications

Boiling Point 120°C (0.5 mmHg)
Quantity 10 g
Formula Weight 192.21
Percent Purity 96%
Chemical Name or Material 7-Acetyl-3,4-dihydro-1,5-benzodioxepin
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'