missing translation for 'onlineSavingsMsg'
Learn More
7-Aminothieno[2,3-b]pyrazine-6-carboxylic acid, ≥90%, Thermo Scientific™ Product Code.: 10641654

7-Aminothieno[2,3-b]pyrazine-6-carboxylic acid, ≥90%, Thermo Scientific™

Product Code. 10641654
1 g, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10641654

Brand: Thermo Scientific Maybridge MO07475DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 56881-31-3
Molecular Formula C7H5N3O2S
Molecular Weight (g/mol) 195.196
MDL Number MFCD08271964
InChI Key UJUCBOIXAMPUQL-UHFFFAOYSA-N
Synonym 7-aminothieno 2,3-b pyrazine-6-carboxylic acid, 7-aminothieno 2,3-b pyrazine-6-carboxylicacid, 7-aminothiopheno 3,2-b pyrazine-6-carboxylic acid, thieno 2,3-b pyrazine-6-carboxylicacid, 7-amino, 7-aminothieno 2,3-b pyrazine-6-carboxylic acid mb41-1
PubChem CID 12225712
IUPAC Name 7-aminothieno[2,3-b]pyrazine-6-carboxylic acid
SMILES C1=CN=C2C(=N1)C(=C(S2)C(=O)O)N

Specifications

CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 195.2
Percent Purity 90%
Chemical Name or Material 7-Aminothieno[2, 3-b]pyrazine-6-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
7-Aminothieno[2,3-b]pyrazine-6-carboxylic acid, ≥90%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.