Learn More
7-Aza-1H-benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate, 98+%, Thermo Scientific™
67.00 € - 111.56 €
Chemical Identifiers
CAS | 156311-85-2 |
---|---|
Molecular Formula | C11H21F6N7OP2 |
Molecular Weight (g/mol) | 443.275 |
MDL Number | MFCD09263289 |
InChI Key | RQBNNDQCKMIUQJ-UHFFFAOYSA-N |
Synonym | 3h-1,2,3 triazolo 4,5-b pyridin-3-yl oxy tris dimethylamino phosphonium hexafluorophosphate v, 7-azabenzotriazol-1-yloxytris dimethylamino phosphonium hexafluorophosphate, 7-azabenzotriazol-1-yloxytris dimethylamino phosph, 7-aza-1h-benzotriazol-1-yloxytris dimethylamino phosphonium hexafluorophosphate, acmc-20alxg, tris dimethylamino 3h-1,2,3-triazolo 4,5-b pyridin-3-yloxy phosphonium hexafluorophosphate, ksc173q2f, 7-azabenzotriazol-1-yloxytris dimethyamino phosphonium hexafluorosphate, 7-azabenzotriazol-1-yloxy tris dimethylamino phosphonium hexafluorophosphate, 7-aza-1h-benzotriazol-1-yloxytris dimethylamino phosphonium hexafluorophosphate, 98+%Show More |
PubChem CID | 21894514 |
IUPAC Name | tris(dimethylamino)-(triazolo[4,5-b]pyridin-3-yloxy)phosphanium;hexafluorophosphate |
SMILES | CN(C)[P+](N(C)C)(N(C)C)ON1C2=C(C=CC=N2)N=N1.F[P-](F)(F)(F)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11352469
|
Thermo Scientific Alfa Aesar
H26609.03 |
1 g | N/A | ||||||
11362469
|
Thermo Scientific Alfa Aesar
H26609.06 |
5 g |
111.56 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
156311-85-2 | |
443.275 | |
RQBNNDQCKMIUQJ-UHFFFAOYSA-N | |
21894514 | |
CN(C)[P+](N(C)C)(N(C)C)ON1C2=C(C=CC=N2)N=N1.F[P-](F)(F)(F)(F)F |
C11H21F6N7OP2 | |
MFCD09263289 | |
3h-1,2,3 triazolo 4,5-b pyridin-3-yl oxy tris dimethylamino phosphonium hexafluorophosphate v, 7-azabenzotriazol-1-yloxytris dimethylamino phosphonium hexafluorophosphate, 7-azabenzotriazol-1-yloxytris dimethylamino phosph, 7-aza-1h-benzotriazol-1-yloxytris dimethylamino phosphonium hexafluorophosphate, acmc-20alxg, tris dimethylamino 3h-1,2,3-triazolo 4,5-b pyridin-3-yloxy phosphonium hexafluorophosphate, ksc173q2f, 7-azabenzotriazol-1-yloxytris dimethyamino phosphonium hexafluorosphate, 7-azabenzotriazol-1-yloxy tris dimethylamino phosphonium hexafluorophosphate, 7-aza-1h-benzotriazol-1-yloxytris dimethylamino phosphonium hexafluorophosphate, 98+%Show More | |
tris(dimethylamino)-(triazolo[4,5-b]pyridin-3-yloxy)phosphanium;hexafluorophosphateShow More |
Specifications
156311-85-2 | |
C11H21F6N7OP2 | |
3h-1,2,3 triazolo 4,5-b pyridin-3-yl oxy tris dimethylamino phosphonium hexafluorophosphate v, 7-azabenzotriazol-1-yloxytris dimethylamino phosphonium hexafluorophosphate, 7-azabenzotriazol-1-yloxytris dimethylamino phosph, 7-aza-1h-benzotriazol-1-yloxytris dimethylamino phosphonium hexafluorophosphate, acmc-20alxg, tris dimethylamino 3h-1,2,3-triazolo 4,5-b pyridin-3-yloxy phosphonium hexafluorophosphate, ksc173q2f, 7-azabenzotriazol-1-yloxytris dimethyamino phosphonium hexafluorosphate, 7-azabenzotriazol-1-yloxy tris dimethylamino phosphonium hexafluorophosphate, 7-aza-1h-benzotriazol-1-yloxytris dimethylamino phosphonium hexafluorophosphate, 98+% | |
CN(C)[P+](N(C)C)(N(C)C)ON1C2=C(C=CC=N2)N=N1.F[P-](F)(F)(F)(F)F | |
443.275 | |
443.27 | |
7-Aza-1H-benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate |
∽180°C (decomposition) | |
MFCD09263289 | |
RQBNNDQCKMIUQJ-UHFFFAOYSA-N | |
tris(dimethylamino)-(triazolo[4,5-b]pyridin-3-yloxy)phosphanium;hexafluorophosphate | |
21894514 | |
≥98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Keep cold
Your input is important to us. Please complete this form to provide feedback related to the content on this product.