missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-4-fluoroindole, 97%, Thermo Scientific™

Product Code. 15430698
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 292636-09-0
Molecular Formula C8H5BrFN
Molecular Weight (g/mol) 214.037
MDL Number MFCD04037874
InChI Key PRXUTIXCOBFMMM-UHFFFAOYSA-N
PubChem CID 4342956
IUPAC Name 7-bromo-4-fluoro-1H-indole
SMILES C1=CC(=C2C(=C1F)C=CN2)Br

Specifications

Quantity 250 mg
Sensitivity Air sensitive
Synonym 7-bromo-4-fluoroindole, 4-fluoro-7-bromoindole, pubchem14798, 1h-indole,7-bromo-4-fluoro, 1h-indole, 7-bromo-4-fluoro, ablock ab-12-9428, 7-bromanyl-4-fluoranyl-1h-indole
Formula Weight 214.04
Percent Purity 97%
Chemical Name or Material 7-Bromo-4-fluoroindole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.