missing translation for 'onlineSavingsMsg'
Learn More

7-Bromoindole, 98%, Thermo Scientific™

Product Code. 10525924
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 51417-51-7
Molecular Formula C8H6BrN
Molecular Weight (g/mol) 196.05
MDL Number MFCD00799492
InChI Key RDSVSEFWZUWZHW-UHFFFAOYSA-N
Synonym 7-bromoindole, 1h-indole, 7-bromo, 7-bromo-indole, 7-bromindol, 7-bromoindol, 7-brom-1h-indole, pubchem7318, acmc-209ktm, ksc269i1f
PubChem CID 2757020
IUPAC Name 7-bromo-1H-indole
SMILES C1=CC2=C(C(=C1)Br)NC=C2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 46°C to 47°C
Color Beige-Yellow or Brown
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 196.05
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 7-Bromoindole
Show More Show Less
compliance-icons
Product Identifier
  • 7-Bromoindole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.