Learn More
7-Bromoisatin, 97%, Thermo Scientific Chemicals
63.80 € - 343.00 €
Chemical Identifiers
CAS | 20780-74-9 |
---|---|
Molecular Formula | C8H4BrNO2 |
Molecular Weight (g/mol) | 226.029 |
MDL Number | MFCD00774354 |
InChI Key | OCVKSIWBTJCXPV-UHFFFAOYSA-N |
Synonym | 7-bromoisatin, 7-bromoindoline-2,3-dione, 7-bromo-2,3-dioxoindoline, 1h-indole-2,3-dione, 7-bromo, 7-bromoindole-2,3-dione, 7-bromo-2,3-dihydro-1h-indole-2,3-dione, 7-bromoindole-1h-2,3-dione, 7-bromo-2,3-indolinedione, 7-bromo isatin, 7-bromo-isatin |
PubChem CID | 2302353 |
IUPAC Name | 7-bromo-1H-indole-2,3-dione |
SMILES | C1=CC2=C(C(=C1)Br)NC(=O)C2=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11394806
|
Thermo Scientific Alfa Aesar
H32843.03 |
1 g |
63.80 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11384806
|
Thermo Scientific Alfa Aesar
H32843.09 |
10 g |
343.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
7-bromoisatin was treated with excess PhMgBr to afford tertiary alcohol 9 in 96 % yield. Isatin oxime triflates undergo a facile fragmentation which is promoted by DBU, to form anthranilonitriles after hydrolytic work-up.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications7-bromoisatin was treated with excess PhMgBr to afford tertiary alcohol 9 in 96 % yield. Isatin oxime triflates undergo a facile fragmentation which is promoted by DBU, to form anthranilonitriles after hydrolytic work-up.
Solubility
Slightly soluble.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
20780-74-9 | |
226.029 | |
OCVKSIWBTJCXPV-UHFFFAOYSA-N | |
2302353 | |
C1=CC2=C(C(=C1)Br)NC(=O)C2=O |
C8H4BrNO2 | |
MFCD00774354 | |
7-bromoisatin, 7-bromoindoline-2,3-dione, 7-bromo-2,3-dioxoindoline, 1h-indole-2,3-dione, 7-bromo, 7-bromoindole-2,3-dione, 7-bromo-2,3-dihydro-1h-indole-2,3-dione, 7-bromoindole-1h-2,3-dione, 7-bromo-2,3-indolinedione, 7-bromo isatin, 7-bromo-isatin | |
7-bromo-1H-indole-2,3-dione |
Specifications
20780-74-9 | |
C8H4BrNO2 | |
7-bromoisatin, 7-bromoindoline-2,3-dione, 7-bromo-2,3-dioxoindoline, 1h-indole-2,3-dione, 7-bromo, 7-bromoindole-2,3-dione, 7-bromo-2,3-dihydro-1h-indole-2,3-dione, 7-bromoindole-1h-2,3-dione, 7-bromo-2,3-indolinedione, 7-bromo isatin, 7-bromo-isatin | |
OCVKSIWBTJCXPV-UHFFFAOYSA-N | |
7-bromo-1H-indole-2,3-dione | |
2302353 | |
97% |
195°C to 199°C | |
MFCD00774354 | |
Slightly soluble. | |
C1=CC2=C(C(=C1)Br)NC(=O)C2=O | |
226.029 | |
226.03 | |
7-Bromoisatin |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.