missing translation for 'onlineSavingsMsg'
Learn More
7-Chloro-1,2,3,4-tetrahydroisoquinoline hydrochloride, 98%, Thermo Scientific Chemicals Product Code.: 15458316

7-Chloro-1,2,3,4-tetrahydroisoquinoline hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 15458316
5 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15458316

Brand: Thermo Scientific Alfa Aesar H63456.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73075-45-3
Molecular Formula C9H11Cl2N
Molecular Weight (g/mol) 204.094
MDL Number MFCD08461076
InChI Key OGIAIXMUSSACDB-UHFFFAOYSA-N
Synonym 7-chloro-1,2,3,4-tetrahydroisoquinoline hydrochloride, 7-chloro-1,2,3,4-tetrahydroisoquinolinehydrochloride, isoquinoline, 7-chloro-1,2,3,4-tetrahydro-, hydrochloride, 7-chloro-1,2,3,4-tetrahydro-isoquinoline hcl, 7-chhloro-1,2,3,4-tetrahydroisoquinoline hydrochloride, 7-chloro-1,2,3,4-tetrahydroisoquinoline hydrochlor, 7-chloro-1,2,3,4-tetrahydroisoquinoline-hydrogen chloride 1/1
PubChem CID 12595070
IUPAC Name 7-chloro-1,2,3,4-tetrahydroisoquinoline;hydrochloride
SMILES C1CNCC2=C1C=CC(=C2)Cl.Cl

Specifications

Quantity 5 g
Formula Weight 204.1
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 7-Chloro-1,2,3,4-tetrahydroisoquinoline hydrochloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW11F093 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberH63456.06,H63456.MD,H63456.03
Lot Number81401120 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberH63456.06,H63456.MD,H63456.03
Lot Number81301249 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberH63456.06,H63456.MD,H63456.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
7-Chloro-1,2,3,4-tetrahydroisoquinoline hydrochloride, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.