missing translation for 'onlineSavingsMsg'
Learn More

7-Deazaguanine, 90+%, Thermo Scientific™

Product Code. 15244814 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15244814

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 7355-55-7
Molecular Formula C6H6N4O
Molecular Weight (g/mol) 150.14
MDL Number MFCD09033259
InChI Key OLAFFPNXVJANFR-UHFFFAOYSA-N
Synonym 7-deazaguanine, 2-amino-3,7-dihydropyrrolo 2,3-d pyrimidin-4-one, 2-amino-7h-pyrrolo 2,3-d pyrimidin-4-ol, 2-amino-4-hydroxypyrrolo 2,3-d pyrimidine, 2-amino-3h-pyrrolo 2,3-d pyrimidin-4 7h-one, unii-gpl8t5zo3m, 2-amino-3,7-dihydro-pyrrolo 2,3-d pyrimidin-4-one, gpl8t5zo3m, 2-amino-1h-pyrrolo 2,3-d pyrimidin-4-ol, 2-amino-1h-pyrrolo 2,3-d pyrimidin-4 7h-one
PubChem CID 96253
IUPAC Name 2-amino-1,7-dihydropyrrolo[2,3-d]pyrimidin-4-one
SMILES NC1=NC(=O)C2=C(NC=C2)N1

Specifications

Melting Point >230.0°C
Assay Percent Range 90% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 150.14
Percent Purity 90+%
Chemical Name or Material 7-Deazaguanine
compliance-icons
Product Identifier
  • 7-Deazaguanine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.