missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals 7-Diethylamino-4-methylcoumarin, 99%
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Melting Point | 70°C to 73°C |
Color | Beige to Purple |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Glass Bottle |
Quantity | 5 g |
Solubility Information | Solubility in water: slightly soluble. Other solubilities: soluble in aqueous acid solutions,soluble in nearly all common organic solvents,slightly soluble in aliphatic hydrocarbons |
Formula Weight | 231.29 |
Show More |
Safety and Handling
Product Identifier
- 7-Diethylamino-4-methylcoumarin
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : 7-Diethylamino-4-methylcoumarin
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'