missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 7-Diethylamino-4-methylcoumarin, 99%

Product Code. 15199163
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-44-1
Molecular Formula C14H17NO2
Molecular Weight (g/mol) 231.29
MDL Number MFCD00006864
InChI Key AFYCEAFSNDLKSX-UHFFFAOYSA-N
Synonym 7-diethylamino-4-methylcoumarin, coumarin 1, 7-diethylamino-4-methyl-2h-chromen-2-one, coumarin 47, coumarin 460, 4-methyl-7-diethylamino coumarin, blancophor aw, blancophor ffg, 4-methyl-7-diethylaminocoumarin, hakkol p
PubChem CID 7050
ChEBI CHEBI:51938
IUPAC Name 7-(diethylamino)-4-methylchromen-2-one
SMILES CCN(CC)C1=CC2=C(C=C1)C(=CC(=O)O2)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 70°C to 73°C
Color Beige to Purple
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 500 g
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in aqueous acid solutions,soluble in nearly all common organic solvents,slightly soluble in aliphatic hydrocarbons
Formula Weight 231.29
Percent Purity 99%
Grade Pure
Physical Form Crystalline Powder
Chemical Name or Material 7-Diethylamino-4-methylcoumarin, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'