missing translation for 'onlineSavingsMsg'
Learn More

7-Iodoisatin, 97%, Thermo Scientific™

Product Code. 15429007
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20780-78-3
Molecular Formula C8H4INO2
Molecular Weight (g/mol) 273.029
MDL Number MFCD01248407
InChI Key RLGIFVDILDEIHL-UHFFFAOYSA-N
Synonym 7-iodoindoline-2,3-dione, 7-iodoisatin, 7-iodo-isatin, 7-iodo-2,3-dihydro-1h-indole-2,3-dione, 7-iodo-indole-2,3-dione, pubchem20658, 7-iodoindole-2,3-dione, 7-iodo-2,3-indolinedione, 7-iodo indole-2,3-dione, 7-iodo indoline-2,3-dione
PubChem CID 4043705
IUPAC Name 7-iodo-1H-indole-2,3-dione
SMILES C1=CC2=C(C(=C1)I)NC(=O)C2=O

Specifications

Quantity 250 mg
Sensitivity Light sensitive
Formula Weight 273.03
Percent Purity 97%
Chemical Name or Material 7-Iodoisatin
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'