missing translation for 'onlineSavingsMsg'
Learn More

7-Methoxycoumarin-4-acetic acid, Thermo Scientific Chemicals

Precursor to coumarin-type fluorescent reagents

47.30 € - 145.00 €

Chemical Identifiers

CAS 62935-72-2
Molecular Formula C12H10O5
Molecular Weight (g/mol) 234.21
MDL Number MFCD00009774
InChI Key ZEKAXIFHLIITGV-UHFFFAOYSA-N
Synonym 7-methoxycoumarin-4-acetic acid, 2-7-methoxy-2-oxo-2h-chromen-4-yl acetic acid, 7-methoxy-2-oxo-2h-chromen-4-yl acetic acid, 7-methoxy-2-oxo-2h-1-benzopyran-4-acetic acid, 2h-1-benzopyran-4-acetic acid, 7-methoxy-2-oxo, 2-7-methoxy-2-oxochromen-4-yl acetic acid, 7-methoxy-2-oxo-2h-chromen-4-yl-acetic acid, 7-methoxycoumarin-4-yl acetic acid, 7-methoxy-2-oxochromen-4-yl acetic acid, mca-oh
PubChem CID 342221
ChEBI CHEBI:51666
IUPAC Name 2-(7-methoxy-2-oxochromen-4-yl)acetic acid
SMILES COC1=CC=C2C(CC(O)=O)=CC(=O)OC2=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15484849
View Documents Promotion Details
Thermo Scientific Alfa Aesar
J62475.MC
100 mg
47.30 €
100mg
Please to purchase this item. Need a web account? Register with us today!
15494849
View Documents Promotion Details
Thermo Scientific Alfa Aesar
J62475.MD
250 mg
73.60 €
250mg
Please to purchase this item. Need a web account? Register with us today!
15474849
View Documents Promotion Details
Thermo Scientific Alfa Aesar
J62475.03
1 g
145.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

7-Methoxycoumarin-4-acetic acid is a precursor to coumarin-type fluorescent reagents. It has been uses as fluorescent probe in preparation of two novel LysB29 selectively labeled fluorescent derivatives of human insulin, in the preparation of cell-penetrating peptide transportan 10 (tp10) and as fluorescent label for peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Methoxycoumarin-4-acetic acid is a precursor to coumarin-type fluorescent reagents. It has been uses as fluorescent probe in preparation of two novel LysB29 selectively labeled fluorescent derivatives of human insulin, in the preparation of cell-penetrating peptide transportan 10 (tp10) and as fluorescent label for peptides.

Solubility
Soluble in acetone and dimethyl sulfoxide.

Notes
Desiccate at -20°C. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

62935-72-2
234.21
ZEKAXIFHLIITGV-UHFFFAOYSA-N
342221
2-(7-methoxy-2-oxochromen-4-yl)acetic acid
C12H10O5
MFCD00009774
7-methoxycoumarin-4-acetic acid, 2-7-methoxy-2-oxo-2h-chromen-4-yl acetic acid, 7-methoxy-2-oxo-2h-chromen-4-yl acetic acid, 7-methoxy-2-oxo-2h-1-benzopyran-4-acetic acid, 2h-1-benzopyran-4-acetic acid, 7-methoxy-2-oxo, 2-7-methoxy-2-oxochromen-4-yl acetic acid, 7-methoxy-2-oxo-2h-chromen-4-yl-acetic acid, 7-methoxycoumarin-4-yl acetic acid, 7-methoxy-2-oxochromen-4-yl acetic acid, mca-oh
CHEBI:51666
COC1=CC=C2C(CC(O)=O)=CC(=O)OC2=C1

Specifications

62935-72-2
Green
C12H10O5
7-methoxycoumarin-4-acetic acid, 2-7-methoxy-2-oxo-2h-chromen-4-yl acetic acid, 7-methoxy-2-oxo-2h-chromen-4-yl acetic acid, 7-methoxy-2-oxo-2h-1-benzopyran-4-acetic acid, 2h-1-benzopyran-4-acetic acid, 7-methoxy-2-oxo, 2-7-methoxy-2-oxochromen-4-yl acetic acid, 7-methoxy-2-oxo-2h-chromen-4-yl-acetic acid, 7-methoxycoumarin-4-yl acetic acid, 7-methoxy-2-oxochromen-4-yl acetic acid, mca-oh
ZEKAXIFHLIITGV-UHFFFAOYSA-N
2-(7-methoxy-2-oxochromen-4-yl)acetic acid
342221
234.21
7-Methoxycoumarin-4-acetic acid
193°C
87°C (189°F)
MFCD00009774
Soluble in acetone and dimethyl sulfoxide.
COC1=CC=C2C(CC(O)=O)=CC(=O)OC2=C1
234.21
CHEBI:51666
Crystalline Solid
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only