missing translation for 'onlineSavingsMsg'
Learn More

7-Methoxycoumarin, 98+%, Thermo Scientific Chemicals

Product Code. 11412797
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

7-Methoxycoumarin is a compound biosynthesized in lavender that exhibits antimicrobial activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Methoxycoumarin is a compound biosynthesized in lavender that exhibits antimicrobial activity.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 531-59-9
Molecular Formula C10H8O3
Molecular Weight (g/mol) 176.171
MDL Number MFCD00006876
InChI Key LIIALPBMIOVAHH-UHFFFAOYSA-N
Synonym 7-methoxycoumarin, herniarin, ayapanin, methylumbelliferone, 7-methoxy-2h-chromen-2-one, herniarine, 7-methoxy-2h-1-benzopyran-2-one, 2h-1-benzopyran-2-one, 7-methoxy, coumarin, 7-methoxy, methoxycourmarin, 7
PubChem CID 10748
ChEBI CHEBI:5679
IUPAC Name 7-methoxychromen-2-one
SMILES COC1=CC2=C(C=C1)C=CC(=O)O2

Specifications

Melting Point 117°C to 120°C
Quantity 5 g
Beilstein 141728
Solubility Information Insoluble in water.
Formula Weight 176.17
Percent Purity ≥98%
Chemical Name or Material 7-Methoxycoumarin

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'