missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ 70-205 Series Yellow Neptune™ Light Weight Gloves

Product Code. 12610972
Click to view available options
Size:
7
8
9
10
This item is not returnable. View return policy
This item is not returnable. View return policy

Designed to provide mechanical strength in light-to-heavy duty handling and production applications. Neptune™ Aramid fiber Industrial Knitted Gloves offer enhanced comfort and dexterity.

  • High-cut resistance with Kevlar™ Aramid fibers reinforcement
  • Neptune coating allows glove fibers to repel liquids
  • Reinforced thumb welt increases abrasion resistance
  • Ambidextrous
  • Washable and reusable
  • Knitwrist cuff
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 420:2003 + A1:2009 and EN 388:2003,EN 407:2004
Classification Standards CATEGORY III EN 388 : 134X, EN 407, ANSI
Description Knit construction
Coating Material Neptune
Color Yellow
Cut Resistance Rating Very high cut resistance
Material Dupont™ Kevlar™ Aramid Fibers
Product Type Mechanical Protection Gloves
Size 8
Thickness (English) 13 gauge
Glove Cuff Style Knit
Liner Material Dupont™ Kevlar™ aramid fibers
Packaging Type 12 pairs in a bag, 12 bags in a carton
Quantity 144 Pairs
Series 70-205
Length (Metric) 220 to 280 mm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'