Learn More
700 mOsm/kg Osmolality Standard, Reagecon
BIOPHARMA
Used to calibrate, control or validate osmometers
Brand:  Reagecon RE-OSM-700
103.72 EUR valid until 2025-02-13
Use promo code "24126" to get your promotional price.
Product Code. 16368185
  • 138.00 € / Pack of 12
Explore more special offers
Description
Description
  • Manufactured in compliance with European, United States and Chinese Pharmacopoeia guidelines
  • Tested by ISO 17025 accredited test method
  • ISO 17025 accreditation for calibration of laboratory balances
  • Prepared gravimetrically on weight/weight basis
Specifications
Specifications
Amber glass ampoule
Osmolality Standard
Calibration, Control or Validate Osmometer
12 x 5 mL
Osmometer