Learn More
Hamilton™ 700 Series Cemented Needle Custom Syringes
Syringes for customized needles. This series of syringes is ideal for dispensing volumes from 0.5 up to 500μL.
Brand:  Hamilton™ 80308
 View more versions of this product
Product Code. 10664301
  • 110.00 € / Each
Explore more special offers
Description
Description
  • The original hand-fitted Hamilton™ syringe
  • Stainless steel plunger is manufactured to fit the glass barrel with a tolerance less than three thousandths of a millimeter
  • Unsurpassed syringe life
Order Info
This is a Custom Order. Please consult with your Fisher Sales Representative before ordering.
Specifications
Specifications
Borosilicate Glass
Syringe
10 μL
6.6 mm
1/Ea.
19.1 mm