missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 7000 Series Copolymer Microsphere Suspensions

Product Code. 11588642 Shop All Thermo Scientific Products
Click to view available options
Diameter (Metric):
3.2 μm
6 μm
7.9 μm
11 μm
12 μm
17 μm
19 μm
25 μm
45 μm
55 μm
71 μm
90 μm
97 μm
134 μm
222 μm
Quantity:
15 mL
100 mL
1000 mL
Unit Size:
1000mL
100mL
15mL
This item is not returnable. View return policy

Product Code. 11588642

Brand: Thermo Scientific™ 7516C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Meet the need for particulate materials with a variety of particle sizes and properties with Thermo Scientific™ 7000 Series Copolymer Microsphere Suspensions.

Meet the need for particulate materials with a variety of particle sizes and properties with Thermo Scientific™ 7000 Series Copolymer Microsphere Suspensions, useful as model systems for fluid mechanics experiments and as challenge particles for large pore filtration systems.

Suspensions of large copolymer microspheres are composed of polystyrene polymer, cross-linked with 4 to 8% divinylbenzene (DVB). The particles are chemically inert and can be washed with alcohol and vacuum or air dried. The polymer density is 1.05g/cm3 and the index of refraction is 1.59 at 589nm. They are packaged as aqueous suspensions at 10% solids by weight.

Ordering Information:

15mL, 100mL and 1000mL bottle sizes available. 15mL (“A” Part No. bottles), 100mL (“B” bottles) and 1000mL (“C” bottles) are packaged to order; example (Part No. 7503B and 7503C). Please allow 10 working days when ordering these fill volumes.

Includes:

Material Safety Data Sheet (MSDS) and general product handling insert sheet

Recommended for:

High concentration polymer particles > 3μm used in filter evaluation and testing, fluid mechanics research, dispersion studies and many other research and development projects. Not intended for use in instrument calibration or diagnostic reagents because they lack the exacting specifications needed for those applications. Also useful as experimental particles for acoustical and optical analytical systems.

TRUSTED_SUSTAINABILITY

Specifications

Additive Contains trace amount of surfactant to inhibit agglomeration and promote stability
Concentration 10%
Content And Storage Unless otherwise stated, refrigerate (2° to 8°C) product when not in use but do not freeze. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer.
Description Particles useful as model systems for fluid mechanics experiments and as challenge particles for large pore filtration systems.
Color White or Colorless
For Use With (Application) Fluid Mechanics, Large Pore Filtration
Material Polystyrene Divinylbenzene (PS-DVB)
Product Type Copolymer Microsphere Suspensions
Includes General product handling insert sheet
Packaging 1000 mL
Particle Density 1.05g/cm3
Quantity 1000 mL
Refractive Index 1.59 at 589nm (25°C)
Stability Product stable for a minimum of 24 months
Diameter (Metric) 17 μm
Show More Show Less

Not intended for use in instrument calibration or diagnostic reagents.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.