Learn More
Hamilton™ 7000 Series Knurled Hub Syringes
Modified Microliter syringes with a plunger-in-needle design.
Brand:  Hamilton™ DS88400
 View more versions of this product
Product Code. 11518044
  • 2010.00 € / Each
Explore more special offers
Description
Description
  • The only zero dead volume syringe available
  • Fine gauge tungsten plunger wire extends to the tip of the needle
  • Ensures that all solvent is expelled