missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ RNase P Instrument Verification Plate for 7300/7500 Systems, 96-well

Used to verify the performance of the Applied Biosystems™ 7300 and 7500 RT-PCR Systems.

Brand:  Applied Biosystems™ 4350584

623.70 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.


Product Code. 10015504

  • 1386.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The 7300 and 7500 Real-Time PCR Systems TaqMan™ RNase P Instrument Verification Plate is a 96-well PCR plate preloaded with the necessary reagents to detect and quantitate genomic copies of the human RNase P gene, a single-copy gene encoding the RNA moiety of the RNase P enzyme. Each well contains preloaded reaction mix (1X TaqMan™ Universal PCR Master Mix, RNase P primers, and FAM™ dye–labeled probe) and template. The 7300 and 7500 Real-Time PCR Systems TaqMan™ RNase P Instrument Verification Plate satisfies the same requirements as the Applied Biosystems™ 7300 and 7500 Real-Time PCR Systems installations. To pass installation, the instruments must demonstrate the ability to distinguish between 5,000 and 10,000 genomic equivalents with a 99.7% confidence level for a subsequent sample run in a single well. To ensure accurate results, Applied Biosystems™ recommends that the performance of your instrument be verified every six months.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Contains one 96-well MicroAmp PCR tray pre-loaded and sealed with RNaseP TaqMan™ reagents, including probes and primers.

Store at -15°C to -25°C. Protect from light.
7300 System, 7500 System
TaqMan™
Real Time PCR (qPCR)
1 kit
qPCR
FAM
Primer-probe
Rnase P Instrument Verification Plate
Dry Ice
RNase P Calibration, Instrument Verification Plate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.