missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell™ 76-301 Series Tiger Paw™ PVC Light Weight Gloves
Made from polyester plated with cotton, Tiger Paw™ offers many features and benefits at an economical price. This heavier construction ensures a more robust glove for heavier applications.
40.05 €
Specifications
Back Material | PVC |
---|---|
Certifications/Compliance | Standard CATEGORY II EN 388 : 214X |
Cuff Material | PVC |
Color | Natural, Blue |
Product Type | Lightweight Gloves |
Description
- Made from polyester plated with cotton
- Knitted construction
- Polyester cotton liner
- Dotted on palm side finish
- Knitwrist cuff
Specifications
PVC | |
PVC | |
Lightweight Gloves | |
Polyester Cotton | |
12 Pairs |
Standard CATEGORY II EN 388 : 214X | |
Natural, Blue | |
Knit | |
Dotted-palm | |
76-301 |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto