missing translation for 'onlineSavingsMsg'
Learn More

Brady™ 7950 Series Thermal Transfer Printer Ribbon, 90 mm Width

Thermal transfer ribbon with high resistance against smearing and chemicals.

Brand:  Brady™ 236004

Product Code. 11548714

  • 125.00 € / 1 box
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Strong environmental resistance against smearing and chemicals.

  • Reliable, sharp-edge print quality maximizes bar code readability
Specifications

Specifications

Black
Wax, Resin
3.5 in.
90 mm
25.4 mm
2011/65/EU and 2015/863
BradyPrinter i7100 Industrial Label Printer, BradyPrinter PR Plus Series Label Printer, THT 76 mm Core Printer
984 ft.
300 m
1 in.
Open-core Ribbon
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers