Learn More
Abnova™ 7q33/CEN7q FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Brand:  Abnova™ FG0195.100uL
Product Code. 16093305
  • 1515.00 € / 100µL
Explore more special offers
Specifications
Store at 4°C in the dark
Fluorescent In Situ Hybridization (Cell)
100 μL
Tube
DNA/RNA