missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ 7X™ Laboratory Detergents
Safe, effective cleaning solutions for glass and plastic labware
104.00 € - 1605.00 €
Specifications
Form | Liquid |
---|---|
pH | 6 to 7.5 |
Concentration | 7X |
Content And Storage | 15°C to 30°C |
For Use With (Application) | Use on Glass, Plastic, benchtop equipment and tubing |
Product Code | Brand | Volume (English) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (English) | Price | Quantity & Availability | |||||
11459575
|
MP Biomedicals™
097667093 |
1 gal. |
104.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12740748
|
MP Biomedicals™
097667094 |
1 gal. |
245.00 €
|
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11479575
|
MP Biomedicals™
097667095 |
5 gal. |
192.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11489575
|
MP Biomedicals™
097667098 |
55 gal. | N/A | ||||||
17194175
|
MP Biomedicals™
097667098 |
55 gal. |
1605.00 €
208.19 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
7X cleaning solutions are ideal for glass and plastic labware. 7X products contain powerful sequestering reagents to prevent deposits that could harm equipment or critical experimental work, and they will not etch delicate glassware or plastic labware. The environmentally friendly, phosphate-free ES formulation is completely soluble in water at any concentration and drains completely from glassware in seconds.
7X Laboratory Detergents
- For manual washing, phosphate-free
- Comprised of concentrated liquids with an anionic-surface active agent and special solvents—compatible with alkalis and acids
- A 5% solution in distilled water has a 6 to 9pH range with alkaline salts added as builders
- For automatic or mechanical washers—effective in temperatures from ambient to 85°C (185°F)
- Highly concentrated, stabilized, and buffered solution with a unique non-ionic surfactant that is stable to acids and bases, phosphate-free
- Contain a powerful wetting agent to clean microscopic crevices not normally accessible to ordinary detergents
- A 1% solution in distilled water has a 9 to 11pH range
Specifications
Liquid | |
7X | |
Use on Glass, Plastic, benchtop equipment and tubing |
6 to 7.5 | |
15°C to 30°C |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 7X Cleaning solution
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : 2-(2-butoxyethoxy)ethanol, Dioctyl Sulfosuccinate Sodium Salt