missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ 7X™-PF-O-Matic Detergent

7X™-PF-O-Matic Detergent is a Phosphate Free detergent, specifically developed for safe and effective cleaning of laboratory instruments and glassware.

149.00 € - 3685.00 €

Specifications

Form Liquid
Appearance Dark Yellow
View More Specs

Products 4
Product Code Brand Volume (English) Price Quantity & Availability  
Product Code Brand Volume (English) Price Quantity & Availability  
11489585
View Documents
MP Biomedicals™
097667594
1 gal.
395.00 €
Pack of 4
Please to purchase this item. Need a web account? Register with us today!
11499585
View Documents
MP Biomedicals™
097667595
5 gal.
546.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
11479585
View Documents
MP Biomedicals™
097667593
1 gal.
149.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
11409595
View Documents
MP Biomedicals™
097667598
55 gal.
3685.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Environmentally safe and phosphate free
  • Developed for safe and effective cleaning of laboratory instruments and glassware including test tubes, petri-dishes, flasks, microscopy slides, pathology equipment and surgical instruments
  • Powerful wetting and penetrating agent, designed to clean under pressure, heat and agitation
  • Behaves and remains mostly suds-free when agitated
  • Several convenient package sizes are available
  • Stable for long-term storage
  • Completely drains from glassware in seconds
  • Eliminates residue leaving no interfering fluorescence
  • pH neutral, and no need for pH adjustment at any concentration
  • No etch to glass or plastic labware at any concentration
Specifications

Specifications

Liquid
Dark Yellow
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • ES-7X-O-MATIC

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H373-May cause damage to organs.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : sodium xylenesulphonate, OCTYLPHENOXYPOLYETHOXY - POLYPROPROXY PROPANOL, Ethylene Glycol, 2,4,7,9-te

SDS
Documents

Documents

Product Certifications