missing translation for 'onlineSavingsMsg'
Learn More
8-Amino-1-naphthol-3,6-disulfonic acid, monosodium salt hydrate, 80%, tech., Thermo Scientific Chemicals Product Code.: 10218803

8-Amino-1-naphthol-3,6-disulfonic acid, monosodium salt hydrate, 80%, tech., Thermo Scientific Chemicals

Product Code. 10218803
500 g, Plastic Bottle
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic Bottle
This item is not returnable. View return policy

Product Code. 10218803

Brand: Thermo Scientific Acros 400845000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5460-09-3
Molecular Formula C10H8NNaO7S2·H2O
Molecular Weight (g/mol) 359.32
MDL Number MFCD00150460
InChI Key QPILZZVXGUNELN-UHFFFAOYSA-M

Specifications

CAS Min % 75.0
CAS Max % 100.0
Color Brown to Yellow
Loss on Drying 4 % to 6 % (1 g, 150°C, 4 hrs)
Infrared Spectrum Authentic
Assay Percent Range 75% min. (HPLC)
Packaging Plastic Bottle
Quantity 500 g
Synonym h acid sodium, 1-amino-8-naphthol-3,6-disulfonic acid monosodiumsalt, 4-amino-5-hydroxynaphthalene-2,7-disulfonic acid 2-sodium salt
Solubility Information Solubility in water: slightly soluble.
SMILES C1=C2C=C(C=C(C2=C(C=C1S(=O)(=O)O)N)[O-])S(=O)(=O)O.[Na+]
IUPAC Name sodium;8-amino-3,6-disulfonaphthalen-1-olate
PubChem CID 102116466
Formula Weight 359.32
Percent Purity 80%
Grade Technical
Physical Form Powder
Chemical Name or Material 8-Amino-1-naphthol-3, 6-disulfonic acid, monosodium salt monohydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
8-Amino-1-naphthol-3,6-disulfonic acid, monosodium salt hydrate, 80%, tech., Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.