missing translation for 'onlineSavingsMsg'
Learn More

8-Bromo-cGMP, sodium salt, Tocris Bioscience™

Product Code. 18094032
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

cGMP analog; activates PKG

Membrane permeable cGMP analog that activates protein kinase G (PKG). 4.3-fold more potent than cGMP in activating PKG1α and promotes relaxation of tracheal and vascular smooth muscle tissue in vitro.

Chemical Identifiers

CAS 51116-01-9
Molecular Formula C10H11BrN5O7P
Molecular Weight (g/mol) 424.10
MDL Number MFCD00070128
InChI Key YUFCOOWNNHGGOD-UMMCILCDSA-N
Synonym 8-bromoguanosine-3',5'-cyclic monophosphate sodium salt
PubChem CID 131879957
IUPAC Name 9-[(4aR,6R,7R,7aS)-2,7-dihydroxy-2-oxo-hexahydro-2λ⁵-furo[3,2-d][1,3,2]dioxaphosphinin-6-yl]-2-amino-8-bromo-6,9-dihydro-3H-purin-6-one
SMILES NC1=NC(=O)C2=C(N1)N([C@@H]1O[C@@H]3COP(O)(=O)O[C@H]3[C@H]1O)C(Br)=N2

Specifications

Quantity 10 mg
Formula Weight 446.09
Percent Purity >98%
Chemical Name or Material 8-Bromo-cGMP, sodium salt
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.