missing translation for 'onlineSavingsMsg'
Learn More
Learn More
8-Bromoguanosine hydrate, 97%, Thermo Scientific Chemicals
22.70 € - 241.00 €
Chemical Identifiers
CAS | 332359-99-6 |
---|---|
Molecular Formula | C10H12BrN5O5 |
Molecular Weight (g/mol) | 362.14 |
MDL Number | MFCD00150531 |
InChI Key | ASUCSHXLTWZYBA-UMMCILCDSA-N |
Synonym | 8-bromoguanosine hydrate, 8-bromoguanosine dihydrate, 2r,3r,4s,5r-2-2-amino-8-bromo-6-hydroxypurin-9-yl-5-hydroxymethyl oxolane-3,4-diol dihydrate, 3s,2r,4r,5r-5-2-amino-8-bromo-6-hydroxypurin-9-yl-2-hydroxymethyl oxolane-3,4-diol, hydrate, hydrate, 2-amino-8-bromo-9-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-1h-purin-6 9h-one dihydrate |
PubChem CID | 18530343 |
IUPAC Name | 2-amino-8-bromo-9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one;dihydrate |
SMILES | NC1=NC2=C(N=C(Br)N2[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)C(=O)N1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11317925
|
Thermo Scientific Alfa Aesar
L02992.03 |
1 g |
22.70 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11327925
|
Thermo Scientific Alfa Aesar
L02992.06 |
5 g |
71.80 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11337925
|
Thermo Scientific Alfa Aesar
L02992.14 |
25 g | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
332359-99-6 | |
362.14 | |
ASUCSHXLTWZYBA-UMMCILCDSA-N | |
18530343 | |
NC1=NC2=C(N=C(Br)N2[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)C(=O)N1 |
C10H12BrN5O5 | |
MFCD00150531 | |
8-bromoguanosine hydrate, 8-bromoguanosine dihydrate, 2r,3r,4s,5r-2-2-amino-8-bromo-6-hydroxypurin-9-yl-5-hydroxymethyl oxolane-3,4-diol dihydrate, 3s,2r,4r,5r-5-2-amino-8-bromo-6-hydroxypurin-9-yl-2-hydroxymethyl oxolane-3,4-diol, hydrate, hydrate, 2-amino-8-bromo-9-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-1h-purin-6 9h-one dihydrate | |
2-amino-8-bromo-9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one;dihydrate |
Specifications
332359-99-6 | |
C10H12BrN5O5 | |
578725 | |
ASUCSHXLTWZYBA-UMMCILCDSA-N | |
NC1=NC2=C(N=C(Br)N2[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)C(=O)N1 | |
362.14 | |
362.14 (Anhydrous) | |
8-Bromoguanosine hydrate |
>300°C | |
MFCD00150531 | |
8-bromoguanosine hydrate, 8-bromoguanosine dihydrate, 2r,3r,4s,5r-2-2-amino-8-bromo-6-hydroxypurin-9-yl-5-hydroxymethyl oxolane-3,4-diol dihydrate, 3s,2r,4r,5r-5-2-amino-8-bromo-6-hydroxypurin-9-yl-2-hydroxymethyl oxolane-3,4-diol, hydrate, hydrate, 2-amino-8-bromo-9-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-1h-purin-6 9h-one dihydrate | |
−30° (c=2 in 1:1 DMSO/Water) | |
2-amino-8-bromo-9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one;dihydrate | |
18530343 | |
97% |
Safety and Handling
missing translation for 'einecsNumber' : 223-677-6
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only