missing translation for 'onlineSavingsMsg'
Learn More
Learn More
8-Bromoquinoline, 97%, Thermo Scientific™
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
10 g
25 g
Specifications
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Density | 1.5940g/mL |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 25 g |
Formula Weight | 208.06 |
Percent Purity | 97% |
Chemical Name or Material | 8-Bromoquinoline |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'