missing translation for 'onlineSavingsMsg'
Learn More
Learn More
8-Hydroxyquinaldine, 98%, Thermo Scientific Chemicals
15.10 € - 102.00 €
Chemical Identifiers
CAS | 826-81-3 |
---|---|
Molecular Formula | C10H9NO |
Molecular Weight (g/mol) | 159.19 |
MDL Number | MFCD00006765 |
InChI Key | NBYLBWHHTUWMER-UHFFFAOYSA-N |
Synonym | 8-hydroxyquinaldine, 2-methyl-8-hydroxyquinoline, 2-methyl-8-quinolinol, 8-quinolinol, 2-methyl, 2-methyloxine, 8-hydroxy-2-methylquinoline, hydroxyquinaldine, 8-hydroxyqinaldine, 2-methyl-quinolin-8-ol, 2-methyl 8-quinolonol |
PubChem CID | 13224 |
IUPAC Name | 2-methylquinolin-8-ol |
SMILES | CC1=NC2=C(C=CC=C2O)C=C1 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
826-81-3 | |
159.19 | |
NBYLBWHHTUWMER-UHFFFAOYSA-N | |
13224 | |
CC1=NC2=C(C=CC=C2O)C=C1 |
C10H9NO | |
MFCD00006765 | |
8-hydroxyquinaldine, 2-methyl-8-hydroxyquinoline, 2-methyl-8-quinolinol, 8-quinolinol, 2-methyl, 2-methyloxine, 8-hydroxy-2-methylquinoline, hydroxyquinaldine, 8-hydroxyqinaldine, 2-methyl-quinolin-8-ol, 2-methyl 8-quinolonol | |
2-methylquinolin-8-ol |
Specifications
826-81-3 | |
100.0 | |
Beige to Brown | |
139°C | |
98% | |
MFCD00006765 | |
8-hydroxyquinaldine, 2-methyl-8-hydroxyquinoline, 2-methyl-8-quinolinol, 8-quinolinol, 2-methyl, 2-methyloxine, 8-hydroxy-2-methylquinoline, hydroxyquinaldine, 8-hydroxyqinaldine, 2-methyl-quinolin-8-ol, 2-methyl 8-quinolonol | |
NBYLBWHHTUWMER-UHFFFAOYSA-N | |
2-methylquinolin-8-ol | |
13224 | |
98% | |
8-Hydroxyquinaldine, 98% |
97.5 | |
70°C to 74°C | |
267°C | |
Authentic | |
C10H9NO | |
21, 106 | |
Solubility in water: insoluble | |
CC1=NC2=C(C=CC=C2O)C=C1 | |
159.19 | |
159.19 | |
Crystalline Powder |
Safety and Handling
missing translation for 'einecsNumber' : 212-562-6
missing translation for 'rtecsNumber' : VC7920000
missing translation for 'tsca' : TSCA
RUO – Research Use Only