missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxyquinoline-2-carboxylic acid, 98%, Thermo Scientific Chemicals

Product Code. 10182961
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1571-30-8
Molecular Formula C10H6NO3
Molecular Weight (g/mol) 188.16
MDL Number MFCD00168972
InChI Key UHBIKXOBLZWFKM-UHFFFAOYSA-M
Synonym 8-hydroxyquinaldic acid, quinaldic acid, 8-hydroxy, 2-quinolinecarboxylic acid, 8-hydroxy, 8-hydroxy-2-quinolinecarboxylic acid, 8hc, acmc-20a4qz, 5-22-05-00272 beilstein handbook reference, 8-hydroxyquinolin-2-carboxylic acid, 8-hydroxyquinoline-2-carbaxylicacid, quinolinecarboxylic acid, 8-hydroxy
PubChem CID 74079
IUPAC Name 8-hydroxyquinoline-2-carboxylic acid
SMILES OC1=C2N=C(C=CC2=CC=C1)C([O-])=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 213.0°C to 214.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Formula Weight 189.17
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 8-Hydroxyquinoline-2-carboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 8-Hydroxyquinoline-2-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'