missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxyquinoline sulfate monohydrate, 98%, Thermo Scientific Chemicals

Product Code. 10237113
2.5 kg, Plastic Bottle
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207386-91-2
Molecular Formula 0·5 H2O
Molecular Weight (g/mol) 203.2
MDL Number MFCD00149610
InChI Key BNCXJZDIJIVJJO-UHFFFAOYSA-N
Synonym 8-quinolinol hemisulfate, 8-quinolinol hemisulfate hemihydrate, 8-hydroxyquinoline hemisulfate salt hemihydrate, quinolin-8-ol; sulfuric acid; hydrate, quinolin-8-ol sulfate hydrate 2:1:1, 8-hydroxyquinoline sulfate, 8-quinolinol sulfate 2:1 monohydrate, 8-quinolinol, sulfate, hydrate 2:1:1, 8-quinolinol hemisulfate salt hemihydrate, pestanal r , analytical standard

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 182.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic Bottle
Quantity 2.5 kg
Merck Index 14, 4843
Solubility Information Solubility in water: soluble in water. Other solubilities: slightly soluble in alcohol, insoluble in ether
Formula Weight 203.2
Percent Purity 98%
Physical Form Crystalline Powder or Lumps
Chemical Name or Material 8-Hydroxyquinoline sulfate monohydrate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 8-Hydroxyquinoline sulfate monohydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
8-Hydroxyquinoline sulfate monohydrate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.