missing translation for 'onlineSavingsMsg'
Learn More
Learn More
8-Methoxypsoralen, 99%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Packaging:
Glass bottle
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Spezifikation
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Melting Point | 143.0°C to 148.0°C |
Color | White to Cream |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (on anhydrous substance) (HPLC) |
Packaging | Glass bottle |
Quantity | 5 g |
Beilstein | 19, I, 711 |
Merck Index | 15, 6059 |
Mehr anzeigen |
Sicherheit und Handhabung
Produktidentifikator
- 8-Methoxypsoralen
- Danger
- Acute toxicity Category 4
- Carcinogenicity Category 1A
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 1A
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H302-Harmful if swallowed.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur