Learn More
8 Regulated Phthalates and BPA in Medium Density Polyethylene, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand:  SPEX Certiprep CRM-PE002
Product Code. 15286026
  • 257.00 € / 5g
Explore more special offers
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
AnalyteCAS No.Concentration
Diisodecyl phthalate26761-40-030,000μg/g
Diisononyl phthalate28553-12-030,000μg/g
Bis(2-Ethylhexyl)phthalate117-81-73000μg/g
Bisphenol A80-05-73000μg/g
Butylbenzyl phthalate85-68-73000μg/g
Diethyl phthalate84-66-23000μg/g
Dimethyl phthalate131-11-33000μg/g
Di-n-butyl phthalate84-74-23000μg/g
Di-n-octyl phthalate117-84-03000μg/g
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
8 Regulated Phthalates and BPA in Medium Density PE
5 g
AA, ICP
Certified Reference Material
2mL Amber Prescored Glass Ampule
Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Polyethylene
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
8 Regulated Phthalates and BPA in Medium Density Polyethylene, SPEX CertiPrep™ > 5g; 2mL amber prescored glass ampule
Product intended for laboratory use only