Learn More
Ibidi 8 Well Sticky-Slide
8 Well Sticky-Slide is an 8 well bottomless μ-Slide with a self-adhesive underside.
Brand:  Ibidi 80828
Encompass
 View more versions of this product
Product Code. 17246213
  • POA / Pack of 15
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
  • An 8 well chamber
  • bottomless with self-adhesive underside
  • sterilized
Specifications
Specifications
Sterile
8
15
Microscope Slide
Low Wall Wells