missing translation for 'onlineSavingsMsg'
Learn More
80% Ethanol, Molecular Biology Grade, Thermo Scientific™ Product Code.: 16622284

80% Ethanol, Molecular Biology Grade, Thermo Scientific™

Product Code. 16622284 Shop All Thermo Scientific Products
4 L, Poly Bottle
Click to view available options
Quantity:
2.5 L
4 L
Packaging:
Glass Bottle
Poly Bottle
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Product Code. 16622284

Brand: Fisher Chemical T08204K7

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Ready made ethanol/water blend prequalified for molecular biology research applications.

  • 80% ethanol and water blend
  • DNase-, RNase-, and protease-free
  • Ready made ethanol/water blend
  • Pre-qualified for molecular biology research applications
  • Suitable for the purification and precipitation of nucleic acids (DNA and RNA) and proteins
  • Tested for use with Applied Biosystems™ MagMAX™ kits
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-17-5, 7732-18-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
IUPAC Name ethanol
SMILES CCO

Specifications

Color Clear, Colorless Liquid
Quantity 4 L
Packaging Poly Bottle
Assay Percent Range 78% to 82%
Residue after Evaporation 0.001% Maximum
Percent Purity 80%
Grade Molecular Biology
Chemical Name or Material Ethyl Alcohol
compliance-icons compliance-icons
Product Identifier
  • 80% Ethanol, Molecular Biology Grade
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : ethanol

For Manufacturing and Laboratory Use. Not for beverage use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
80% Ethanol, Molecular Biology Grade, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.