missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ 80 Position Collection Plate

Collection Plate for 1.5 mL Vials

Brand:  Gilson™ 12040102

Product Code. 16686105

  • 23.85 € / Each
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Purification Technique is HPLC
  • Maximum Vessel Quantity is 80
  • Vessel Type is tubes
Specifications

Specifications

1.5 mL Vial
80
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers