missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ 80 Position Fraction Collection Plate

These accessories are for use with Gilson fraction collectors.

Brand:  Gilson™ 12240102

 View more versions of this product

Product Code. 16606115

  • 244.00 € / Pack of 20
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Includes drop detector heads, dispense needles, collection plates, and a multiple column adapter
  • Vessel Type is Cavities
  • Purification Technique is HPLC
Specifications

Specifications

2.5 mL Cavities
20 Pk.
80
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.