Learn More
Gilson™ 80 Position Rack
These racks are for use with the FC203B and FC204 Fraction Collectors.
Brand:  Gilson™ 12240101
Product Code. 16696105
  • 171.72 € / Pack of 6
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • HPLC Purification Technique
  • Fraction Collector Racks
Specifications
Specifications
Code 1
13 x 65/100 mm Vial
80