missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek 800 TS Absorbance Reader
GREENER_CHOICE

The BioTek 800 TS absorbance reader is an affordable, high-quality microplate reader for assays in 6- to 384-well formats. For Research Use Only. Not for use in diagnostic procedures.

Brand:  Agilent BioTek 800TSUV-SN

5532.80 EUR valid until 2025-12-31
Use promo code "24302" to get your promotional price.


 View more versions of this product

Product Code. 17277799

  • 5965.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The BioTek 800 TS absorbance reader is an affordable, high-quality microplate reader for assays in 6- to 384-well formats. Applications include ELISA, protein quantification, enzyme kinetics and cell-based assays

The color touch screen provides a visual user interface, making programming fast and intuitive. Expanded functionality and data analysis is available with Gen5 software. The 800 TS partners with the 50 TS washer, making a compact, versatile, and affordable system to automate many application workflows.

Services

12 months

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Microplate Reader
USB Type B, USB Type A
Endpoint, Kinetic, Well-area Scanning
Gen5 Data Analysis Software, Gen5 Secure for 21 CFR Part 11 compliance (option)
CE and TUV marked. RoHS compliant.
Measuring absorbance in microplate-based assays
800TSUV
340 to 750 nm
8 kg
Photodiode
Tungsten Halogen Lamp
Linear (except NB configurations)
10 nm
340, 405, 450, 490, 630 nm
6- to 96-well plates; 340 to 750 nm; Shaking
100/240 V
18.5 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Agilent BioTek 800 TS Absorbance Reader > 6- to 96-well plates; 340 to 750nm wavelength; Linear shaking

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.