missing translation for 'onlineSavingsMsg'
Learn More

LMS™ Series 1A Model 80NP Cooled Incubator, 76 L, Pre-coated Aluminum

Temperature Range: -10°C to +50°C

Brand:  LMS™ 80NP

Product Code. 11873410

  • 5470.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Range of cooled incubators with a white stove enamel steel exterior, foam insulation and a white pre-coated aluminum interior
  • Reliable and accurate temperature control
  • Standard and programmable models available
  • Programmable models offer multiple temperature control in multiple time zones
  • Temperature range -10.0°C to +50.0°C, temperature variation with average workload ±0.5°C
  • Self-evaporating condensate drip tray
  • White plastic coated wire shelves
  • Energy efficient and eco-friendly
  • P.I.D. heating and cooling to ensure stable internal temperature
  • Pre-set safety cutout
  • Hermetically sealed refrigeration system
  • Door lock
  • Controls are easy to read and simple to use
  • Full range of optional extras available. including LED lighting, access porthole, inner perspex doors, alarm system, internal socket and chart recorder
  • Ideal for applications such as growth studies, B.O.D.tests, seed germination and vernalisation, tissue culture, enzymology, serum studies and immunology, sample storage or product shelf life tests
Specifications

Specifications

ISO 9001:2008
Benchtop or underbench incubator
Pre-coated Aluminum
420 mm x 386 mm
Stove Enameled Mild Steel
-10°C to +50°C
Digital
Foamed polyurethane
CFC-free
2 Years
17.5 in.
445 mm
15.7 in.
400 mm
490 mm
240 V
Programmable
76 L
Fan assisted air circulation via premixing chamber
Optional LED
2
Refrigerated Incubator
2 trays
80NP
±0.1°C
20.5 in.
520 mm
30.7 in.
780 mm
19.3 in.
40 kg
50/60 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers