missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 850 Homogenizer Accessory, Motor Stand Assembly
GREENER_CHOICE

Fisher Scientific™ 850 Homogenizer Accessory, Motor Stand Assembly designed to retain the Fisher Scientific 850 Homogenizer for post mounted operation.

Brand:  Fisherbrand™ 15535819

216.06 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.


Product Code. 15535819

  • 240.00 € / Each
Estimated Shipment: 16-05-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Includes a baseplate and post; does not include motor clamp
  • Base dimensions: 12.7 x 20.3cm (5 x 8 in.) stainless steel base
  • Post dimensions: 48.2cm (19 in.) tall by 9.52mm (0.375 in.) diameter stainless steel support post
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Motor Stand Assembly
Stainless Steel
Fisher Scientific 850 Homogenizer
Baseplate and post
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ 850 Homogenizer Accessory, Motor Stand Assembly > Motor Stand Assembly

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.