Learn More
Tuttnauer 85L Benchtop Autoclave
The 3870EL-D has an advanced multi-color control panel and a chamber made of 316L stainless steel. Each model has a number of optional added-value features which can be configured for fast cooling, efficient drying, biohazard and waste sterilization, Fo control, and more.
Brand: Tuttnauer 3870EL-230-D
Description
Designed for your laboratory applications. The 3870EL-D has an advanced multi-color control panel and a chamber made of 316L stainless steel. Each model has a number of optional added-value features which can be configured for fast cooling, efficient drying, Fo control, and more.
- Advanced Control System - Take Advantage of Tuttnauer’s state-of-the-art control system with multi-color display
- Safe - Automatic pressure locking system. Door will not open when the chamber is overpressured
- Optional Internal Printing - Documents date, time, temperature and pressure
- Intuitive Multi-Color Display - Multi-color display for easier reading gives quick access to important information. Color is used to indicate the stage of the cycle
- High Quality Stainless Steel - 316L grade stainless steel chamber and door with electro-polish finish
- Diverse Load Types - The 3870EL-D autoclave comes with 8 factory set cycle programs that cover the processing needs of most laboratories and research centers
Specifications
18.7 gal. | |
70 cm | |
28 in. | |
37.6 in. | |
Benchtop Autoclave | |
105°C to 137°C | |
316L Stainless Steel |
85 L | |
15.2 in. | |
38 cm | |
94 cm | |
221°F to 279°F | |
Single Phase | |
Non-sterile |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.