missing translation for 'onlineSavingsMsg'
Learn More

Tuttnauer 85L Vertical Autoclave

The 3870ELV-D has an advanced multi-color control panel and a chamber made of 316L stainless steel. Each model has a number of optional added-value features which can be configured for fast cooling, efficient drying, biohazard and waste sterilization, Fo control, and more.

Brand:  Tuttnauer 3870ELV-230-D

 View more versions of this product

Product Code. 17841077

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Features

  • Fo software control
  • PID (Proportional Integral Differential) pressure control
  • Stores the last 200 cycles in built-in memory
  • Two independent flexible PT100 temperature sensors to prevent over-boiling of liquids and explosions of bottles
  • Up to 6 temperature sensors and 4 pressure sensors can be connected
  • 30 Identification Codes and Passwords for access level control
  • The controller and software comply with the 21 CFR part 11 standard

Innovative Multi-Color Display

  • Multi-color display for easier reading
  • Color is used to indicate the stage of the cycle
  • Quick access to important information
  • 26 Languages
  • Built-in view of historical cycle data
Specifications

Specifications

18.7 gal.
70 cm
28 in.
31.6 in.
Autoclave, Top-loading
105°C to 138°C
316L Stainless Steel
Non-sterile
85 L
15.2 in.
38 cm
54 cm
221°F to 280°F
Single Phase, 230 V
230 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers