missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 8mm PP Screw Seal, Pink, Center hole, 8-425 thread, Assembled septum

Polypropylene caps are chemically inert and suitable for most chromatography applications.

Brand:  Fisherbrand™ 08 15 2493

 View more versions of this product

Product Code. 13475499

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers

Description

Description

  • 8mm
  • PP
  • Screw Cap
  • Pink
  • Center hole
  • 8-425 thread
  • Pre-assembled caps and septa are convenient and minimize contamination from handling
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Silicone/PTFE
8 mm
Yes
100
Screw Cap
Red, White, Pink
Polypropylene
GC, HPLC
1.3 mm
8-425
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers