missing translation for 'onlineSavingsMsg'
Learn More

9,10-Dimethylanthracene, 97%, Thermo Scientific Chemicals

Product Code. 11430627
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

9,10-Dimethylanthracene is used as an antioxidant-photosensitizer dual-loaded polymeric micelles with controllable production of reactive oxygen species. It is also used as chemical rate constant actinometer in singlet molecular oxygen reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9,10-Dimethylanthracene is used as an antioxidant-photosensitizer dual-loaded polymeric micelles with controllable production of reactive oxygen species. It is also used as chemical rate constant actinometer in singlet molecular oxygen reactions.

Solubility
Soluble in toluene. Insoluble in water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 781-43-1
Molecular Formula C16H14
Molecular Weight (g/mol) 206.29
MDL Number MFCD00001262
InChI Key JTGMTYWYUZDRBK-UHFFFAOYSA-N
Synonym anthracene, 9,10-dimethyl, unii-di986077vr, ccris 2742, 9,10-dimethyl-anthracene, gnf-pf-2329, acmc-209pdj, 9,10-dimethylanthracen, 9:10-dimethylanthracene, anthracene,10-dimethyl, 9,10-dimethylanthraceneShow MoreShow Less
PubChem CID 13076
IUPAC Name 9,10-dimethylanthracene
SMILES CC1=C2C=CC=CC2=C(C)C2=CC=CC=C12

Specifications

Melting Point 182°C to 184°C
Quantity 1 g
Beilstein 1909028
Solubility Information Soluble in toluene. Insoluble in water.
Formula Weight 206.29
Percent Purity 99%
Chemical Name or Material 9,10-Dimethylanthracene

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10242521 Certificate TypeCertificate of Analysis Date04/08/2022 Catalog NumberB22458.06,B22458.03
Lot Number10239911 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberB22458.06,B22458.03
Lot Number10229034 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22458.06,B22458.03
Lot Number10235006 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22458.06,B22458.03
Lot Number10214806 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22458.06,B22458.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
9,10-Dimethylanthracene, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.