Learn More
9,10-Diphenylanthracene, 99%, Thermo Scientific Chemicals
Description
9,10-Diphenylanthracene acts as a sensitizer in chemiluminescence. It is used in light sticks in order to produce blue light. It is also useful in blue organic light-emitting diodes (OLEDs) and OLED-based displays. Further, it plays an important role in the preparation of 9,10-difluoro-9,10-diphenyl-9,10-dihydro-anthracene. In addition to this, it is used as an organic semiconductor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
9,10-Diphenylanthracene acts as a sensitizer in chemiluminescence. It is used in light sticks in order to produce blue light. It is also useful in blue organic light-emitting diodes (OLEDs) and OLED-based displays. Further, it plays an important role in the preparation of 9,10-difluoro-9,10-diphenyl-9,10-dihydro-anthracene. In addition to this, it is used as an organic semiconductor.
Solubility
Soluble in ethanol, chloroform, benzene and carbon disulfide. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.

Specifications
Specifications
Melting Point | 248°C to 254°C |
Odor | Odorless |
Quantity | 25 g |
Beilstein | 1914010 |
Solubility Information | Soluble in ethanol,chloroform,benzene and carbon disulfide. Insoluble in water. |
Formula Weight | 330.43 |
Percent Purity | 99% |
Chemical Name or Material | 9,10-Diphenylanthracene |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
S16I039 | Certificate of Analysis | 22/06/2022 | A15714.03,A15714.14,A15714.06 |
R31F018 | Certificate of Analysis | 12/04/2022 | A15714.03,A15714.14,A15714.06 |
P24A034 | Certificate of Analysis | 23/11/2021 | A15714.03,A15714.14,A15714.06 |
L20W037 | Certificate of Analysis | 23/11/2021 | A15714.03,A15714.14,A15714.06 |
90009194 | Certificate of Analysis | 23/11/2021 | A15714.03,A15714.14,A15714.06 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.