Learn More
9,9-Dimethyl-4,5-bis(diphenylphosphino)xanthene, 98%, Thermo Scientific Chemicals
50.15 € - 823.10 €
Chemical Identifiers
CAS | 161265-03-8 |
---|---|
Molecular Formula | C39H32OP2 |
Molecular Weight (g/mol) | 578.63 |
MDL Number | MFCD00233866 |
InChI Key | CXNIUSPIQKWYAI-UHFFFAOYSA-N |
Synonym | xantphos, 4,5-bis diphenylphosphino-9,9-dimethylxanthene, 9,9-dimethyl-4,5-bis diphenylphosphino xanthene, 9,9-dimethyl-9h-xanthene-4,5-diyl bis diphenylphosphine, dimethylbisdiphenylphosphinoxanthene, unii-nmu72mog9b, nmu72mog9b, 9,9-dimethyl-9h-xanthene-4,5-diyl bis diphenylphosphane, 5-diphenylphosphanyl-9,9-dimethylxanthen-4-yl-diphenylphosphane, 4,5-bis-diphenylphosphanyl-9,9-dimethyl-9h-xanthene |
PubChem CID | 636044 |
SMILES | CC1(C)C2=C(OC3=C1C=CC=C3P(C1=CC=CC=C1)C1=CC=CC=C1)C(=CC=C2)P(C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10024762
|
Thermo Scientific Acros
378060010 |
1 g | Glass bottle |
50.15 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10397432
|
Thermo Scientific Acros
378060050 |
5 g | Glass bottle |
168.84 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10533094
|
Thermo Scientific Acros
378060250 |
25 g | Glass bottle |
823.10 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
161265-03-8 | |
578.63 | |
CXNIUSPIQKWYAI-UHFFFAOYSA-N | |
636044 |
C39H32OP2 | |
MFCD00233866 | |
xantphos, 4,5-bis diphenylphosphino-9,9-dimethylxanthene, 9,9-dimethyl-4,5-bis diphenylphosphino xanthene, 9,9-dimethyl-9h-xanthene-4,5-diyl bis diphenylphosphine, dimethylbisdiphenylphosphinoxanthene, unii-nmu72mog9b, nmu72mog9b, 9,9-dimethyl-9h-xanthene-4,5-diyl bis diphenylphosphane, 5-diphenylphosphanyl-9,9-dimethylxanthen-4-yl-diphenylphosphane, 4,5-bis-diphenylphosphanyl-9,9-dimethyl-9h-xanthene | |
CC1(C)C2=C(OC3=C1C=CC=C3P(C1=CC=CC=C1)C1=CC=CC=C1)C(=CC=C2)P(C1=CC=CC=C1)C1=CC=CC=C1 |
Specifications
161265-03-8 | |
100.0 | |
White to Yellow | |
98% | |
MFCD00233866 | |
xantphos, 4,5-bis diphenylphosphino-9,9-dimethylxanthene, 9,9-dimethyl-4,5-bis diphenylphosphino xanthene, 9,9-dimethyl-9h-xanthene-4,5-diyl bis diphenylphosphine, dimethylbisdiphenylphosphinoxanthene, unii-nmu72mog9b, nmu72mog9b, 9,9-dimethyl-9h-xanthene-4,5-diyl bis diphenylphosphane, 5-diphenylphosphanyl-9,9-dimethylxanthen-4-yl-diphenylphosphane, 4,5-bis-diphenylphosphanyl-9,9-dimethyl-9h-xanthene | |
CXNIUSPIQKWYAI-UHFFFAOYSA-N | |
(5-diphenylphosphanyl-9,9-dimethylxanthen-4-yl)-diphenylphosphane | |
636044 | |
98% | |
9, 9-Dimethyl-4, 5-bis(diphenylphosphino)xanthene |
97.5 | |
226.0°C to 230.0°C | |
Authentic | |
C39H32OP2 | |
15, 10268 | |
Solubility in water: insoluble | |
CC1(C)C2=C(OC3=C1C=CC=C3P(C1=CC=CC=C1)C1=CC=CC=C1)C(=CC=C2)P(C1=CC=CC=C1)C1=CC=CC=C1 | |
578.63 | |
578.63 | |
Crystals |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Harmful if inhaled.
Harmful if swallowed.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
- 9,9-Dimethyl-4,5-bis(diphenylphosphino)xanthene
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.