missing translation for 'onlineSavingsMsg'
Learn More
9-Anthracenecarboxaldehyde, 99%, Thermo Scientific Chemicals Product Code.: 10754831

9-Anthracenecarboxaldehyde, 99%, Thermo Scientific Chemicals

Product Code. 10754831
100 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10754831

Brand: Thermo Scientific Acros 153931000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 642-31-9
Molecular Formula C15H10O
Molecular Weight (g/mol) 206.24
MDL Number MFCD00001254
InChI Key YMNKUHIVVMFOFO-UHFFFAOYSA-N
Synonym 9-anthraldehyde, 9-anthracenecarboxaldehyde, 9-anthrylaldehyde, 9-formylanthracene, 9-anthracenecarbaldehyde, 9-anthrylcarboxaldehyde, ccris 3165, nsc 15, anthracene-9-aldehyde, anthracene-9-carboxaldehydeShow MoreShow Less
PubChem CID 69504
IUPAC Name anthracene-9-carbaldehyde
SMILES O=CC1=C2C=CC=CC2=CC2=CC=CC=C12

Specifications

Melting Point 103.0°C to 107.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 100 g
Solubility Information Solubility in water: .. Other solubilities: soluble in chloroform
Formula Weight 206.24
Percent Purity 99%
Physical Form Crystalline Powder or Needles
Chemical Name or Material 9-Anthracenecarboxaldehyde
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0456164 Certificate TypeCertificate of Analysis Date08/09/2023 Catalog Number153935000,153931000,153930250
Lot NumberA0452602 Certificate TypeCertificate of Analysis Date05/05/2023 Catalog Number153935000,153931000,153930250
Lot NumberA0395649 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number153935000,153931000,153930250
Lot NumberA0415808 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number153935000,153931000,153930250
Lot NumberA0435556 Certificate TypeCertificate of Analysis Date01/11/2021 Catalog Number153935000,153931000,153930250
Product Identifier
  • 9-Anthracenecarboxaldehyde

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
9-Anthracenecarboxaldehyde, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.