missing translation for 'onlineSavingsMsg'
Learn More

9-Borabicyclo[3.3.1]nonane, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals

77.20 € - 345.00 €

Specifications

Chemical Name or Material 9-Borabicyclo[3.3.1]nonane
CAS 280-64-8, 109-99-9
CAS Min % 6.14, 92.49
CAS Max % 7.51, 93.86
Density 0.8840g/mL
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10315092
View Documents
Thermo Scientific Acros
165811000
100 mL AcroSeal™ Glass bottle
77.20 €
100mL
Please to purchase this item. Need a web account? Register with us today!
10609834
View Documents
Thermo Scientific Acros
165818000
800 mL AcroSeal™ Glass Bottle
345.00 €
800mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid, Reduction reagent
Specifications

Specifications

9-Borabicyclo[3.3.1]nonane
6.14, 92.49
0.8840g/mL
C8H15B
02,31; 03,24; 04,41; 05,46; 06,62; 07,29; 08,47; 09,57; 10,48; 11,68; 13,92; 14,52; 15,43; 17,49; 01,732; 04,356
9-borabicyclo 3.3.1 nonane, 9-borabicyclo 3.3.1 nonan, 9-borabicyclo 3,3,1 nonane, 9$l^ 2-borabicyclo 3.3.1 nonane, 1s,5s-9-borabicyclo 3.3.1 nonane, monomer; 21205-91-4 dimer, 9-borabicyclo 3.3.1 nonane,9-bbn,intetrahydrofuran
AMKGKYQBASDDJB-UHFFFAOYSA-N
9$l^{2}-borabicyclo[3.3.1]nonane
6327450
280-64-8, 109-99-9
7.51, 93.86
−17°C
MFCD00074742
0.884
Solubility in water: reacts
[B]1C2CCCC1CCC2
122.02
122.02
Videos
Safety and Handling

Safety and Handling

GHS H Statement
In contact with water releases flammable gases.
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Suspected of causing cancer.
May form explo

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Handle under inert gas.
Protect from moisture

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 206-000-9

Product Identifier
  • 9-Borabicyclo¢3.3.1!nonane, 0.5M solution in THF

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Substance which\, in contact with water\, emits flammable gases Category 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Carcinogenicity Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H261-In contact with water releases flammable gases.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • EUH019-May form explosive peroxides.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Tetrahydrofuran, 9-Borabicyclo¢3.3.1!nonane

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only