Learn More
9-Bromo-1-nonanol, 97%, Thermo Scientific Chemicals
51.20 € - 162.00 €
Chemical Identifiers
CAS | 55362-80-6 |
---|---|
Molecular Formula | C9H19BrO |
Molecular Weight (g/mol) | 223.15 |
MDL Number | MFCD00063348 |
InChI Key | USJDOLXCPFASNV-UHFFFAOYSA-N |
Synonym | 9-bromo-1-nonanol, 1-nonanol, 9-bromo, 9-bromo-nonan-1-ol, nonamethylene bromohydrin, 9-bromnonan-1-ol, 9-bromononanol, 9-bromononyl alcohol, pubchem23429, 9-bromo nonanol, acmc-1axfq |
PubChem CID | 108700 |
IUPAC Name | 9-bromononan-1-ol |
SMILES | OCCCCCCCCCBr |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
55362-80-6 | |
223.15 | |
USJDOLXCPFASNV-UHFFFAOYSA-N | |
108700 | |
OCCCCCCCCCBr |
C9H19BrO | |
MFCD00063348 | |
9-bromo-1-nonanol, 1-nonanol, 9-bromo, 9-bromo-nonan-1-ol, nonamethylene bromohydrin, 9-bromnonan-1-ol, 9-bromononanol, 9-bromononyl alcohol, pubchem23429, 9-bromo nonanol, acmc-1axfq | |
9-bromononan-1-ol |
Specifications
55362-80-6 | |
125.0°C to 126.0°C (2.0 Torr) | |
Authentic | |
C9H19BrO | |
9-bromo-1-nonanol, 1-nonanol, 9-bromo, 9-bromo-nonan-1-ol, nonamethylene bromohydrin, 9-bromnonan-1-ol, 9-bromononanol, 9-bromononyl alcohol, pubchem23429, 9-bromo nonanol, acmc-1axfq | |
OCCCCCCCCCBr | |
223.15 | |
223.16 | |
9-Bromo-1-nonanol |
30.0°C to 33.0°C | |
109°C | |
96% min. (GC) | |
MFCD00063348 | |
USJDOLXCPFASNV-UHFFFAOYSA-N | |
9-bromononan-1-ol | |
108700 | |
97% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
- 9-Bromo-1-nonanol
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.